5-Amino-1-methyl-1,2-dihydropyridin-2-one 95%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-1-methylpyridin-2(1H)-one

A112511-100MG A112511-250MG A112511-1G A112511-5G
103798-026EA 73 USD
103798-026 103823-862 76970-916 76970-918 77310-672 77310-674
5-Amino-1-methyl-1,2-dihydropyridin-2-one 95%
5-Amino-1-methyl-1,2-dihydropyridin-2-one

Precaución: for Research Use Only

Formula: C₆H₈N₂O
MW: 124.141 g/mol
Boiling Pt: 254.5 °C (760 mm Hg)
Melting Pt: 125…126 °C
Flash Pt: 107.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09814480
CAS Number: 33630-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR