2-(3-Amino-1H-pyrazol-5-yl)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-1H-pyrazole-3-acetic acid

A164860-100MG A164860-250MG A164860-5G A164860-1G
103805-912EA 37 USD
103805-912 103844-170 77138-284 77138-286 77300-736 77320-364
2-(3-Amino-1H-pyrazol-5-yl)acetic acid 97%
2-(3-Amino-1H-pyrazol-5-yl)acetic acid

Precaución: for Research Use Only

Formula: C₅H₇N₃O₂
MW: 141.13 g/mol
Boiling Pt: 504.4 °C (760 mm Hg)
Flash Pt: 258.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09991896
CAS Number: 174891-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR