5-Amino-1H-pyrazol-3(2H)-one 98%

Supplier: Ambeed
Warning

A246130-100G A246130-25G A246130-10G
103851-518EA 13 USD
103851-518 77031-370 77304-376 77668-008
5-Amino-1H-pyrazol-3(2H)-one 98%
5-Amino-1H-pyrazol-3(2H)-one

Precaución: for Research Use Only

Formula: C₃H₅N₃O
Boiling Pt: 479.5 °C (760 mm Hg)
Melting Pt: 204 °C
Flash Pt: 243.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00204482
CAS Number: 28491-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR