5-Amino-1H-imidazole-4-carboxamide hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 5(4)-Amino-4(5)-imidazolecarboxamide Hydrochloride, AICA, 5(4)-Amino-4(5)-(aminocarbonyl)imidazole Hydrochloride, 4-Aminoimidazole-5-carboxamide hydrochloride, 4-Amino-5-imidazolecarboxamide hydrochloride, 5-Amino-4-imidazolecarboxamide hydrochloride

A224063-25G A224063-5G A224063-100G A224063-10G A224063-500G
103849-970EA 6 USD
103849-970 77088-602 77088-604 77275-570 77275-572 77275-574
5-Amino-1H-imidazole-4-carboxamide hydrochloride 98%
5-Amino-1H-imidazole-4-carboxamide hydrochloride

Precaución: for Research Use Only

Formula: C₄H₇ClN₄O
MW: 162.58 g/mol
Boiling Pt: 522.1 °C (760 mm Hg)
Melting Pt: 250…252 °C
Flash Pt: 269.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00012704
CAS Number: 72-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR