5-Amino-1-(pyridin-2-yl)-1H-pyrazole-4-carbonitrile 95%

Supplier: Ambeed
Warning

A101306-5G A101306-250MG
103795-898EA 18 USD
103795-898 103821-584 77297-050 77610-744
5-Amino-1-(pyridin-2-yl)-1H-pyrazole-4-carbonitrile 95%
5-Amino-1-(pyridin-2-yl)-1H-pyrazole-4-carbonitrile

Precaución: for Research Use Only

Formula: C₉H₇N₅
MW: 185.185 g/mol
Boiling Pt: 439.0±35.0 °C (760 mm Hg)
Flash Pt: 219.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00128357
CAS Number: 72816-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR