5-Acetyl-2-methylpyridine 97%

Supplier: Ambeed
Warning

A125783-250MG A125783-5G
103800-722EA 18 USD
103800-722 77094-738 77298-086
5-Acetyl-2-methylpyridine 97%
5-Acetyl-2-methylpyridine

Precaución: for Research Use Only

Formula: C₈H₉NO
Boiling Pt: 233.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00128113
CAS Number: 36357-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR