5-Acetyl-2-methoxypyridine 98%

Supplier: Ambeed
Warning

A114453-100G A114453-25G A114453-1G A114453-5G
103798-406EA 10 USD
103798-406 103824-244 77136-180 77136-182 77311-056 77311-058
5-Acetyl-2-methoxypyridine 98%
5-Acetyl-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MW: 151.1600 g/mol
Boiling Pt: 269.5 °C (760 mm Hg)
Melting Pt: 53…57 °C
Flash Pt: 116.791 °C
Storage Temperature: Ambient
MDL Number: MFCD05664350
CAS Number: 213193-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR