5-Acetyl-2-bromopyridine 97%

Supplier: Ambeed
Warning

Synonyms: 3-Acetyl-6-bromopyridine

A171418-100G A171418-10G A171418-250MG A171418-1G A171418-25G A171418-5G
103806-632EA 6 USD
103806-632 103844-906 77011-582 77011-584 77011-586 77082-060 77082-062 77082-064
5-Acetyl-2-bromopyridine 97%
5-Acetyl-2-bromopyridine

Precaución: for Research Use Only

Formula: C₇H₆BrNO
MW: 200.03 g/mol
Melting Pt: 124…128 °C
Flash Pt: 138.1 °C
Storage Temperature: Ambient
MDL Number: MFCD04974527
CAS Number: 139042-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR