5,8-Dibromoisoquinoline 98%

Supplier: Ambeed
Warning

A100869-10G A100869-250MG A100869-25G A100869-1G A100869-5G
103795-662EA 27 USD
103795-662 103821-354 76765-140 76765-142 76765-144 77296-866 77296-868
5,8-Dibromoisoquinoline 98%
5,8-Dibromoisoquinoline

Precaución: for Research Use Only

Formula: C₉H₅Br₂N
Boiling Pt: 363.7 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD03109888
CAS Number: 81045-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR