5,8-Dibromoimidazo[1,2-a]pyrazine 97%

Supplier: Ambeed
Warning

A193652-100MG A193652-250MG A193652-5G
103816-558EA 99 USD
103816-558 76974-932 76974-934 76974-936
5,8-Dibromoimidazo[1,2-a]pyrazine 97%
5,8-Dibromoimidazo[1,2-a]pyrazine

Precaución: for Research Use Only

Formula: C₆H₃Br₂N₃
Storage Temperature: Refrigerator
MDL Number: MFCD13193554
CAS Number: 957344-74-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR