5,7-Di-tert-butyl-3-(3,4-dimethylphenyl)benzofuran-2(3H)-one 98%

Supplier: Ambeed
Warning

A643193-10G A643193-25G A643193-5G
103840-360EA 19 USD
103840-360 77267-078 77308-050 77308-052
5,7-Di-tert-butyl-3-(3,4-dimethylphenyl)benzofuran-2(3H)-one 98%
5,7-Di-tert-butyl-3-(3,4-dimethylphenyl)benzofuran-2(3H)-one

Precaución: for Research Use Only

Formula: C₂₄H₃₀O₂
MW: 350.50 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30729989
CAS Number: 164391-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR