5,7-Dimethylpyrazolo[1,5-a]pyrimidine-3-carboxylic acid 98%

Supplier: Ambeed
Warning

A188020-10G A188020-250MG A188020-1G A188020-5G
103816-064EA 36 USD
103816-064 103846-654 77029-716 77029-718 77084-372 77084-374
5,7-Dimethylpyrazolo[1,5-a]pyrimidine-3-carboxylic acid 98%
5,7-Dimethylpyrazolo[1,5-a]pyrimidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₉N₃O₂
MW: 191.187 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01051014
CAS Number: 90349-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR