5,7-Dimethylisatin ≥97% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: 5,7-Dimethylindoline-2,3-dione, 5,7-Dimethyl-1H-indole-2,3-dione

A204326-1G A204326-25G A204326-5G A204326-10G
103817-542EA 6 USD
103817-542 103848-204 77302-902 77302-904 77302-906 77567-958
5,7-Dimethylisatin ≥97% (by HPLC)
5,7-Dimethylisatin

Precaución: For research use only.

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Melting Pt: 244…248 °C
Storage Temperature: Ambient
MDL Number: MFCD00047219
CAS Number: 39603-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR