5,7-Dimethoxyindan-1-one 95%

Supplier: Ambeed
Warning

A149204-100MG A149204-250MG A149204-1G A149204-5G
103804-018EA 56 USD
103804-018 103842-138 76768-086 76768-088 77299-714 77299-716
5,7-Dimethoxyindan-1-one 95%
5,7-Dimethoxyindan-1-one

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₃
MW: 192.21 g/mol
Boiling Pt: 352.6 °C (760 mm Hg)
Flash Pt: 158.3 °C
Storage Temperature: Ambient
MDL Number: MFCD02684971
CAS Number: 880-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR