5,7-Difluoro-1-indanone 95%

Supplier: Ambeed
Warning

A211622-10G A211622-5G A211622-1G A211622-25G
103818-126EA 25 USD
103818-126 103848-820 76975-568 77087-240 77303-204 77303-206
5,7-Difluoro-1-indanone 95%
5,7-Difluoro-1-indanone

Precaución: for Research Use Only

Formula: C₉H₆F₂O
MW: 168.14 g/mol
Boiling Pt: 252 °C (760 mm Hg)
Melting Pt: 80…84 °C
Flash Pt: 96 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09908161
CAS Number: 84315-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR