5,7-Dichloro-4-quinolinol 98%

Supplier: Ambeed
Warning

Synonyms: 5,7-Dichloro-4-hydroxyquinoline, 5,7-Dichloroquinolin-4-ol

A142550-100MG A142550-250MG A142550-1G A142550-5G
103803-180EA 55 USD
103803-180 76767-874 76767-876 77316-406 77316-408
5,7-Dichloro-4-quinolinol 98%
5,7-Dichloro-4-quinolinol

Precaución: for Research Use Only

Formula: C₉H₅Cl₂NO
MW: 214.0481 g/mol
Boiling Pt: 340.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00219834
CAS Number: 171850-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR