5,7-Dichloropyrazolo[1,5-a]pyrimidine-3-carbonitrile 97%

Supplier: Ambeed
Warning

A130127-250MG A130127-1G A130127-5G
103801-380EA 34 USD
103801-380 76870-682 77298-390 77298-392
5,7-Dichloropyrazolo[1,5-a]pyrimidine-3-carbonitrile 97%
5,7-Dichloropyrazolo[1,5-a]pyrimidine-3-carbonitrile

Precaución: for Research Use Only

Formula: C₇H₂Cl₂N₄
MW: 213.02 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17392854
CAS Number: 845895-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR