5,7-Dichloropyrazolo[1,5-a]pyrimidine 97%

Supplier: Ambeed
Warning

A171329-1G A171329-25G A171329-5G A171329-100MG A171329-10G A171329-250MG
103806-626EA 16 USD
103806-626 103844-900 77082-048 77082-050 77082-052 77163-578 77163-580 77163-582
5,7-Dichloropyrazolo[1,5-a]pyrimidine 97%
5,7-Dichloropyrazolo[1,5-a]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₃Cl₂N₃
MW: 188.0141 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08273918
CAS Number: 57489-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR