5,7-Dichloro-2-methylthiazolo[5,4-d]pyrimidine ≥95%

Supplier: Ambeed
Warning

A295001-100MG A295001-250MG
103814-036EA 914 USD
103814-036 77885-779 77885-780
5,7-Dichloro-2-methylthiazolo[5,4-d]pyrimidine ≥95%
5,7-Dichloro-2-methylthiazolo[5,4-d]pyrimidine

Precaución: Inert atmosphere, 2…8 °C.

Formula: C₆H₃Cl₂N₃S
Storage Temperature: Refrigerator
MDL Number: MFCD12828095
CAS Number: 7464-11-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR