5,7-Dichloro-1H-imidazo[4,5-b]pyridine 97%

Supplier: Ambeed
Warning

A274049-100MG A274049-10G A274049-250MG A274049-1G A274049-25G A274049-5G
103813-176EA 48 USD
103813-176 103853-100 76787-246 76787-248 76787-250 77092-916 77092-918 77305-156
5,7-Dichloro-1H-imidazo[4,5-b]pyridine 97%
5,7-Dichloro-1H-imidazo[4,5-b]pyridine

Precaución: for Research Use Only

Formula: C₆H₃Cl₂N₃
MW: 188.01 g/mol
Boiling Pt: 416.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD11111407
CAS Number: 24485-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR