5,7-Dichloro-1,6-naphthyridine 97%

Supplier: Ambeed
Warning

A207957-100MG A207957-10G A207957-250MG A207957-1G A207957-25G A207957-5G
103817-842EA 90 USD
103817-842 103848-528 76785-594 76785-596 76785-598 77086-824 77086-826 77086-828
5,7-Dichloro-1,6-naphthyridine 97%
5,7-Dichloro-1,6-naphthyridine

Precaución: for Research Use Only

Formula: C₈H₄Cl₂N₂
MW: 199.0368 g/mol
Boiling Pt: 344.5 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD11518965
CAS Number: 337958-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR