6-Amino-2,3-dimethylpyridine 97%

Supplier: Ambeed
Warning

A382927-25G A382927-10G A382927-250MG
103826-724EA 37 USD
103826-724 103837-760 76841-082 77251-918 77251-920
6-Amino-2,3-dimethylpyridine 97%
6-Amino-2,3-dimethylpyridine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂
Boiling Pt: 231.8 °C (760 mm Hg)
Flash Pt: 116.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06200777
CAS Number: 57963-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR