5,6-Dimethylpyrazine-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A287583-100MG A287583-250MG A287583-1G A287583-5G
103813-766EA 90 USD
103813-766 103834-908 76827-654 76827-656 77305-520 77305-522
5,6-Dimethylpyrazine-2-carboxylic acid 95%
5,6-Dimethylpyrazine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03196496
CAS Number: 13515-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR