5,6-Dimethoxybenzo[d]thiazole 95%

Supplier: Ambeed
Warning

A113739-100MG A113739-250MG A113739-1G
103798-284EA 305 USD
103798-284 103824-128 76982-856 76982-858 77310-942
5,6-Dimethoxybenzo[d]thiazole 95%
5,6-Dimethoxybenzo[d]thiazole

Precaución: for Research Use Only

Formula: C₉H₉NO₂S
MW: 195.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14584602
CAS Number: 58249-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR