5,6-Dimethoxy-2-(4-piperidinylmethyl)-1-indanone hydrochloride 95%

Supplier: Ambeed
Warning

A167420-25G A167420-5G
103844-446EA 9 USD
103844-446 77081-462 77081-464
5,6-Dimethoxy-2-(4-piperidinylmethyl)-1-indanone hydrochloride 95%
5,6-Dimethoxy-2-(4-piperidinylmethyl)-1-indanone hydrochloride

Precaución: for Research Use Only

Formula: C₁₇H₂₄ClNO₃
MW: 325.8304 g/mol
Boiling Pt: 479.4 °C (760 mm Hg)
Melting Pt: 249…250 °C
Storage Temperature: Ambient
MDL Number: MFCD09033179
CAS Number: 120013-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR