5,6-Dihydroxyindole 95%

Supplier: Ambeed
Danger

A172524-250MG A172524-1G A172524-100MG A172524-10G A172524-25G
103806-728EA 55 USD
103806-728 103845-012 76998-272 77082-206 77567-536 77671-452 77671-454
5,6-Dihydroxyindole 95%
5,6-Dihydroxyindole

Precaución: For research use only.

Formula: C₈H₇NO₂
MW: 149.149 g/mol
Boiling Pt: 411.2 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD00798933
CAS Number: 3131-52-0
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR