5,6-Dihydroxy-1H-indole-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A244439-100MG A244439-250MG A244439-5G A244439-1G
103811-580EA 164 USD
103811-580 103851-412 76826-802 76826-804 76826-806 77304-318
5,6-Dihydroxy-1H-indole-2-carboxylic acid 95%
5,6-Dihydroxy-1H-indole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₇NO₄
MW: 193.16 g/mol
Boiling Pt: 578.6 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD02181103
CAS Number: 4790-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR