5,6-Dihydro-4H-pyrrolo[1,2-b]pyrazole-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A195062-100MG A195062-250MG A195062-1G A195062-5G
103816-716EA 95 USD
103816-716 103847-330 76975-008 76975-010 77085-280 77302-440
5,6-Dihydro-4H-pyrrolo[1,2-b]pyrazole-3-carboxylic acid 97%
5,6-Dihydro-4H-pyrrolo[1,2-b]pyrazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
MW: 152.151 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846937
CAS Number: 796729-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR