5,6-Dihydro-4H-pyrrolo[1,2-b]pyrazole-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A215692-100MG A215692-10G A215692-250MG A215692-1G A215692-25G A215692-5G
103818-510EA 18 USD
103818-510 103849-218 76975-670 76975-672 76975-674 77087-762 77087-764 77087-766
5,6-Dihydro-4H-pyrrolo[1,2-b]pyrazole-2-carboxylic acid 97%
5,6-Dihydro-4H-pyrrolo[1,2-b]pyrazole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
Boiling Pt: 381.6 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD11846936
CAS Number: 796729-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR