5,6-Dihydro-2H-pyran-3-carbaldehyde 95%

Supplier: Ambeed
Warning

A362460-10G A362460-5G A362460-250MG
103826-320EA 109 USD
103826-320 103837-336 76773-356 77306-740 77626-702
5,6-Dihydro-2H-pyran-3-carbaldehyde 95%
5,6-Dihydro-2H-pyran-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₆H₈O₂
Boiling Pt: 201.9 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD08146636
CAS Number: 13417-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR