5,6-Difluoro-2,3-dihydro-1H-inden-1-one 97%

Supplier: Ambeed
Warning

A260202-1G A260202-25G A260202-5G A260202-100MG A260202-10G A260202-250MG
103812-484EA 12 USD
103812-484 103852-366 77091-944 77091-946 77091-948 77098-816 77098-818 77098-820
5,6-Difluoro-2,3-dihydro-1H-inden-1-one 97%
5,6-Difluoro-2,3-dihydro-1H-inden-1-one

Precaución: for Research Use Only

Formula: C₉H₆F₂O
MW: 168.14 g/mol
Boiling Pt: 251.3 °C (760 mm Hg)
Flash Pt: 95.65 °C
Storage Temperature: Ambient
MDL Number: MFCD07772125
CAS Number: 161712-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR