5,6-Dichloropyridine-2,3-diamine 95%

Supplier: Ambeed
Warning

A264610-100MG A264610-250MG A264610-5G
103812-686EA 103 USD
103812-686 76816-072 76816-074 77672-370
5,6-Dichloropyridine-2,3-diamine 95%
5,6-Dichloropyridine-2,3-diamine

Precaución: for Research Use Only

Formula: C₅H₅Cl₂N₃
MW: 178.02 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13181782
CAS Number: 97941-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR