5,6-Dichloropyridazin-3(2H)-one 98%

Supplier: Ambeed
Warning

A208957-10G A208957-250MG A208957-1G A208957-5G
103817-904EA 14 USD
103817-904 103848-592 76825-808 76825-810 77086-928 77086-930
5,6-Dichloropyridazin-3(2H)-one 98%
5,6-Dichloropyridazin-3(2H)-one

Precaución: for Research Use Only

Formula: C₄H₂Cl₂N₂O
Boiling Pt: 378 °C (760 mm Hg)
Flash Pt: 182.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04971420
CAS Number: 17285-36-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR