5,6-Dichloronicotinonitrile 95%
Supplier: Ambeed
|
Warning
|
A284435-100MG
A284435-1G
A284435-5G
103813-640EA
36
USD
103813-640
103834-772
77002-240
77305-458
77305-460
5,6-Dichloronicotinonitrile 95%
5,6-Dichloronicotinonitrile
Precaución: for Research Use Only
Formula:
C₆H₂Cl₂N₂ Melting Pt: 146…148 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD11100247 CAS Number: 65189-15-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...