5,6-Dichloro-2-methylbenzimidazole 98%

Supplier: Ambeed
Warning

A135921-250MG A135921-1G A135921-5G
103802-242EA 15 USD
103802-242 76767-492 77315-122 77315-124
5,6-Dichloro-2-methylbenzimidazole 98%
5,6-Dichloro-2-methylbenzimidazole

Precaución: for Research Use Only

Formula: C₈H₆Cl₂N₂
MW: 201.0500 g/mol
Boiling Pt: 409 °C (760 mm Hg)
Melting Pt: 244…245 °C
Storage Temperature: Ambient
MDL Number: MFCD00276973
CAS Number: 6478-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR