5,6-Dichloro-2,3-dihydro-1H-inden-1-one 98%

Supplier: Ambeed
Warning

Synonyms: 5,6-Dichloro-indanone

A109861-250MG A109861-1G A109861-5G A109861-10G
103797-582EA 20 USD
103797-582 103823-386 76970-672 77310-190 77310-192 77670-556
5,6-Dichloro-2,3-dihydro-1H-inden-1-one 98%
5,6-Dichloro-2,3-dihydro-1H-inden-1-one

Precaución: for Research Use Only

Formula: C₉H₆Cl₂O
MW: 201.0493 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09032944
CAS Number: 68755-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR