5,6-Dichloro-1H-pyrrolo[3,2-b]pyridin-2(3H)-one 97%

Supplier: Ambeed
Warning

A186813-100MG A186813-5G
103815-966EA 415 USD
103815-966 103846-554 76985-942 77084-242
5,6-Dichloro-1H-pyrrolo[3,2-b]pyridin-2(3H)-one 97%
5,6-Dichloro-1H-pyrrolo[3,2-b]pyridin-2(3H)-one

Precaución: for Research Use Only

Formula: C₇H₄Cl₂N₂O
MW: 203.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847072
CAS Number: 136888-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR