5,6-Dibromo-2-pyridinamine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5,6-Dibromopyridin-2-amine
A135341-100G
A135341-100MG
A135341-10G
A135341-250MG
A135341-25G
A135341-1G
A135341-5G
103802-138EA
32
USD
103802-138
76983-954
76983-956
76983-958
76983-960
76983-962
77314-952
77314-954
5,6-Dibromo-2-pyridinamine 97%
5,6-Dibromo-2-pyridinamine
Precaución: for Research Use Only
Formula:
C₅H₄Br₂N₂ MW: 251.907 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00234045 CAS Number: 89284-11-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...