5,6-Dibromopyrazin-2-amine 98%

Supplier: Ambeed
Warning

A158337-100MG A158337-250MG
103805-122EA 69 USD
103805-122 77011-090 77011-092
5,6-Dibromopyrazin-2-amine 98%
5,6-Dibromopyrazin-2-amine

Precaución: for Research Use Only

Formula: C₄H₃Br₂N₃
MW: 252.9 g/mol
Boiling Pt: 321.9±37.0 °C (760 mm Hg)
Flash Pt: 148.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD27923208
CAS Number: 1353100-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR