5,6-Dibromopicolinic acid 95%

Supplier: Ambeed
Warning

A111089-100MG A111089-250MG
103797-794EA 76 USD
103797-794 77148-698 77148-700
5,6-Dibromopicolinic acid 95%
5,6-Dibromopicolinic acid

Precaución: for Research Use Only

Formula: C₆H₃Br₂NO₂
MW: 280.9 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11978042
CAS Number: 1206679-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR