5,6-Dibromopyridine-3-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5,6-Dibromonicotinic acid

A183701-1G A183701-5G A183701-25G
103815-660EA 19 USD
103815-660 103846-222 77301-846 77301-848 77671-594
5,6-Dibromopyridine-3-carboxylic acid 98%
5,6-Dibromopyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₃Br₂NO₂
MW: 280.91 g/mol
Boiling Pt: 379.5 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD01927099
CAS Number: 29241-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR