5,6-Dibromo-1,3-benzodioxole 97%
Supplier: Ambeed
|
Warning
|
A142251-1G
A142251-5G
103803-132EA
5
USD
103803-132
76996-908
77316-348
77316-350
5,6-Dibromo-1,3-benzodioxole 97%
5,6-Dibromo-1,3-benzodioxole
Precaución: for Research Use Only
Formula:
C₇H₄Br₂O₂ Boiling Pt: 294.7 °C (760 mm Hg) Melting Pt: 83…85 °C Flash Pt: 118.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00094476 CAS Number: 5279-32-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...