5,6-Diaminopyrimidine-2,4-diol 95%

Supplier: Ambeed
Warning

Synonyms: 5,6-Diaminopyrimidine-2,4-diol, 5,6-Diaminouracil hydrochloride

A268662-250MG A268662-1G
103812-926EA 34 USD
103812-926 103852-828 77001-074 77305-010
5,6-Diaminopyrimidine-2,4-diol 95%
5,6-Diaminopyrimidine-2,4-diol

Precaución: for Research Use Only

Formula: C₄H₆N₄O₂
MW: 142.12 g/mol
Melting Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD00016632
CAS Number: 3240-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR