5,6-Diaminopyrimidin-4(3H)-one sulfate(2:1) 95%

Supplier: Ambeed
Warning

A171243-1G A171243-10G A171243-250MG
103806-612EA 52 USD
103806-612 103844-886 77082-028 77286-864 77286-866
5,6-Diaminopyrimidin-4(3H)-one sulfate(2:1) 95%
5,6-Diaminopyrimidin-4(3H)-one sulfate(2:1)

Precaución: for Research Use Only

Formula: C₈H₁₄N₈O₆S
MW: 350.31 g/mol
Melting Pt: 270 °C
Storage Temperature: Ambient
MDL Number: MFCD17216010
CAS Number: 102783-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR