5,6-Diamino-1,10-phenanthroline 96%

Supplier: Ambeed
Warning

A387443-1G A387443-5G A387443-100MG A387443-250MG A387443-25G
103826-828EA 49 USD
103826-828 103837-870 76841-236 76841-238 76978-894 76978-896 77882-034
5,6-Diamino-1,10-phenanthroline 96%
5,6-Diamino-1,10-phenanthroline

Precaución: For research use only

Formula: C₁₂H₁₀N₄
Boiling Pt: 445.7 °C (760 mm Hg)
Melting Pt: 258…262 °C
Flash Pt: 283.784 °C
Storage Temperature: Ambient
MDL Number: MFCD12913573
CAS Number: 168646-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR