5,6,7,8-Tetrahydroquinoline 97%

Supplier: Ambeed
Warning

A371016-25G A371016-5G A371016-1000G A371016-100G A371016-10G A371016-500G
103837-486EA 5 USD
103837-486 76840-730 76840-732 77033-280 77033-282 77033-284 77033-286
5,6,7,8-Tetrahydroquinoline 97%
5,6,7,8-Tetrahydroquinoline

Precaución: for Research Use Only

Formula: C₉H₁₁N
MW: 133.19 g/mol
Boiling Pt: 222.7 °C (760 mm Hg)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00006734
CAS Number: 10500-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR