5,6,7,8-Tetrahydroquinolin-8-ol 97%
Supplier: Ambeed
|
Warning
|
A183801-1G
A183801-250MG
103815-672EA
13
USD
103815-672
103846-234
77083-834
77151-702
5,6,7,8-Tetrahydroquinolin-8-ol 97%
5,6,7,8-Tetrahydroquinolin-8-ol
Precaución: for Research Use Only
Formula:
C₉H₁₁NO MW: 149.1897 g/mol Boiling Pt: 309.5 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD07369987 CAS Number: 14631-46-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...