5,6,7,8-Tetrahydroquinolin-8-amine 96%

Supplier: Ambeed
Warning

A267773-100MG A267773-250MG
103812-880EA 329 USD
103812-880 103852-780 77166-354 77166-356
5,6,7,8-Tetrahydroquinolin-8-amine 96%
5,6,7,8-Tetrahydroquinolin-8-amine

Precaución: for Research Use Only

Formula: C₉H₁₂N₂
MW: 148.205 g/mol
Boiling Pt: 277.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08234982
CAS Number: 298181-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR