5,6,7,8-Tetrahydroquinolin-3-ol 95%

Supplier: Ambeed
Warning

A299323-10G A299323-250MG
103814-224EA 212 USD
103814-224 103835-370 76988-760 76988-762
5,6,7,8-Tetrahydroquinolin-3-ol 95%
5,6,7,8-Tetrahydroquinolin-3-ol

Precaución: for Research Use Only

Formula: C₉H₁₁NO
MW: 149.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13178913
CAS Number: 655239-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR