5,6,7,8-Tetrahydronaphthalene-1-carboxylic acid 98%

Supplier: Ambeed
Warning

A229934-100G A229934-10G A229934-1G A229934-5G A229934-25G
103819-504EA 7 USD
103819-504 103850-406 76801-068 76801-070 77089-260 77089-262 77303-858
5,6,7,8-Tetrahydronaphthalene-1-carboxylic acid 98%
5,6,7,8-Tetrahydronaphthalene-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₂
MW: 176.2118 g/mol
Flash Pt: 154.2 °C
Storage Temperature: Ambient
MDL Number: MFCD01464030
CAS Number: 4242-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR